Please login to your online account to display your discounted pricing

Tetrapropylammonium perchlorate, 98%, ACROS Organics™


Quantity 100g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  AC420741000

 View more versions of this product

Catalog No. AC420741000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5% (on dry substance)
Physical Form Chunks
Formula Weight 285.8
Quantity 100g
Color White
Linear Formula (CH3CH2CH2)4NClO4
Health Hazard 1 GHS Signal Word: Danger
MDL Number MFCD00041979
Molecular Formula C12H28ClNO4
Chemical Name or Material Tetrapropylammonium perchlorate
Assay Percent Range 98%
CAS 15780-02-6
Packaging Plastic bottle
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
May intensify fire; oxidizer.
Water 2 to 10%
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Tetrapropylammonium perchlorate
Assay ≥97.5% (on dry substance)
Assay Percent Range 98%
Molecular Formula C12H28ClNO4
Linear Formula (CH3CH2CH2)4NClO4
Formula Weight 285.8
MDL Number MFCD00041979
Accessories

GHS Signal Word: Danger

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
May intensify fire; oxidizer.

SDS & Safety Documents