Learn More
Tetrapropylammonium perchlorate, 98%, ACROS Organics™
$177.42 - $177.42
Chemical Identifiers
CAS | 15780-02-6 |
---|---|
Molecular Formula | C12H28ClNO4 |
Molecular Weight (g/mol) | 285.8 |
MDL Number | MFCD00041979 |
InChI Key | LJDKWXHIGFOFDB-UHFFFAOYSA-M |
Synonym | 1-propanaminium, n,n,n-tripropyl-, perchlorate, 1-propanaminium, n,n,n-tripropyl-, perchlorate 1:1, n,n,n-tripropyl-1-propanaminium perchlorate, n,n,n-tripropylpropan-1-aminium perchlorate, tetrapropyl ammonium perchlorate, tetrapropylammonium perchlorate, tetrapropylazanium perchlorate |
PubChem CID | 85095 |
IUPAC Name | tetrapropylazanium;perchlorate |
SMILES | CCC[N+](CCC)(CCC)CCC.[O-]Cl(=O)(=O)=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC420740250
|
Thermo Fisher Scientific
420740250 |
25g | Plastic bottle |
Each for $177.42
|
|
AC420741000
|
Thermo Fisher Scientific
AC420741000 |
100g | Plastic bottle |
N/A
|
N/A |
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Oxidation reagent, Phase transfer catalystChemical Identifiers
15780-02-6 | |
285.8 | |
LJDKWXHIGFOFDB-UHFFFAOYSA-M | |
85095 | |
CCC[N+](CCC)(CCC)CCC.[O-]Cl(=O)(=O)=O |
C12H28ClNO4 | |
MFCD00041979 | |
1-propanaminium, n,n,n-tripropyl-, perchlorate, 1-propanaminium, n,n,n-tripropyl-, perchlorate 1:1, n,n,n-tripropyl-1-propanaminium perchlorate, n,n,n-tripropylpropan-1-aminium perchlorate, tetrapropyl ammonium perchlorate, tetrapropylammonium perchlorate, tetrapropylazanium perchlorate | |
tetrapropylazanium;perchlorate |
Specifications
Authentic | |
2 to 10% | |
238°C to 240°C | |
15780-02-6 | |
C12H28ClNO4 | |
MFCD00041979 | |
LJDKWXHIGFOFDB-UHFFFAOYSA-M | |
tetrapropylazanium;perchlorate | |
85095 | |
Chunks | |
Tetrapropylammonium perchlorate |
Plastic bottle | |
White | |
25g | |
97.5% min. (on dry substance) | |
(CH3CH2CH2)4NClO4 | |
1-propanaminium, n,n,n-tripropyl-, perchlorate, 1-propanaminium, n,n,n-tripropyl-, perchlorate 1:1, n,n,n-tripropyl-1-propanaminium perchlorate, n,n,n-tripropylpropan-1-aminium perchlorate, tetrapropyl ammonium perchlorate, tetrapropylammonium perchlorate, tetrapropylazanium perchlorate | |
CCC[N+](CCC)(CCC)CCC.[O-]Cl(=O)(=O)=O | |
285.8 | |
285.8 | |
98% |
Safety and Handling
GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
May intensify fire; oxidizer.
GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 239-874-5