Please login to your online account to display your discounted pricing

Tetrapropylammonium iodide, 98+%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  216690250

 View more versions of this product

Catalog No. AC216690250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.0%
Physical Form Crystalline Powder or Crystals
Formula Weight 313.26
Quantity 25g
Color White to slightly yellow
TSCA TSCA
EINECS Number 211-157-1
RTECS Number BS8410000
Beilstein 04, 140
Infrared Spectrum Authentic
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material Tetrapropylammonium iodide
Name Note 98+%
Assay Percent Range 98+%
Molecular Formula C12H28IN
Linear Formula (CH3CH2CH2)4NI
MDL Number MFCD00011842
CAS 631-40-3
Health Hazard 1 HYGROSCOPIC
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Tetrapropylammonium iodide
Name Note 98+%
CAS 631-40-3
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C12H28IN
Linear Formula (CH3CH2CH2)4NI
Formula Weight 313.26
MDL Number MFCD00011842
Accessories

HYGROSCOPIC

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents