Please login to your online account to display your discounted pricing

Tetrapropylammonium iodide, 98+%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Tetrapropylammonium iodide
Name Note 98+%
CAS 631-40-3
Assay 98.0% min.
Assay Percent Range 98+%
Molecular Formula C12H28IN
Linear Formula (CH3CH2CH2)4NI
Formula Weight 313.26g/mol
MDL Number MFCD00011842
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC216690250

  SDS
thermo fisher scientific
216690250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Infrared Spectrum Authentic
Packaging Glass bottle
Color White to slightly yellow
Quantity 25g
Chemical Name or Material Tetrapropylammonium iodide
Name Note 98+%
CAS 631-40-3
Physical Form White to slightly yellow crystalline powder or crystals
Assay 98.0% min.
Assay Percent Range 98+%
Molecular Formula C12H28IN
Linear Formula (CH3CH2CH2)4NI
Formula Weight 313.26g/mol
MDL Number MFCD00011842
Beilstein 04, 140
Health Hazard 1 HYGROSCOPIC
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
RTECS Number BS8410000
EINECS Number 211-157-1
TSCA TSCA

Phase transfer catalyst

Safety and Handling

HYGROSCOPIC

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.