Please login to your online account to display your discounted pricing

Tetrapropylammonium hydroxide, 25% in water, ACROS Organics™


Quantity 500g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  AC420735000

 View more versions of this product

Catalog No. AC420735000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 203.37
Quantity 500g
TSCA TSCA
EINECS Number 224-800-6
RTECS Number BS8400000
Density 1g/mL
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 GHS Signal Word: Danger
CAS 7732-18-5
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Molecular Formula C12H29NO
Packaging Plastic bottle
Name Note 25% in Water
Chemical Name or Material Tetrapropylammonium hydroxide
CAS 4499-86-9
Linear Formula (CH3CH2CH2)4NOH
MDL Number MFCD00009360

Specifications

Chemical Name or Material Tetrapropylammonium hydroxide
Name Note 25% in Water
CAS 4499-86-9
CAS 7732-18-5
Molecular Formula C12H29NO
Linear Formula (CH3CH2CH2)4NOH
Formula Weight 203.37
MDL Number MFCD00009360
Accessories

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.

SDS & Safety Documents