Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetraphenyltin, 97%

Chemical Identifiers

Chemical Name or Material Tetraphenyltin
CAS 595-90-4
Molecular Formula (C6H5)4Sn
Formula Weight 427.11
MDL Number MFCD00003006
UN Number UN3146
Synonym Tetraphenylstannane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1913218

  SDS
alfa aesar™
A1913218
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 427.11
Chemical Name or Material Tetraphenyltin
Boiling Point >420°C
TSCA TSCA
Melting Point 224° to 227°C
Assay 97%
Molecular Formula (C6H5)4Sn
MDL Number MFCD00003006
CAS 595-90-4
Quantity 50g
UN Number UN3146
Synonym Tetraphenylstannane
Flash Point >110°C (230°F)
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Odor Characteristic
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, SOLID, N.O.S.
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P361-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.