Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetraphenylthiophene, 97%

$84.04 - $350.96

Chemical Identifiers

Chemical Name or Material Tetraphenylthiophene
CAS 1884-68-0
Molecular Formula C28H20S
Formula Weight 388.53
MDL Number MFCD00014527
Beilstein 297707
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2258803

 
alfa aesar™
B2258803
1g Each for $84.04

AAB2258806

 
alfa aesar™
B2258806
5g Each for $350.96
Description & Specifications

Specifications

Formula Weight 388.53
Chemical Name or Material Tetraphenylthiophene
Assay 97%
Molecular Formula C28H20S
MDL Number MFCD00014527
CAS 1884-68-0
Beilstein 297707
Melting Point 178° to 186°C
Quantity 1g