Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetraphenylsilane, 96%

$250.12 - $250.12

Chemical Identifiers

Chemical Name or Material Tetraphenylsilane
CAS 1048-08-4
Molecular Formula C24H20Si
Formula Weight 336.51
MDL Number MFCD00014069
Beilstein 1885911
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0485714

  SDS
alfa aesar™
L0485714
25g Each for $250.12
Description & Specifications

Specifications

Formula Weight 336.51
Chemical Name or Material Tetraphenylsilane
MDL Number MFCD00014069
CAS 1048-08-4
Molecular Formula C24H20Si
Quantity 25g
TSCA TSCA
Boiling Point 228°C (3mmHg)
Beilstein 1885911
Melting Point 235° to 239°C
Flash Point 193°C (379°F)
Assay 96%

Applications
Tetraphenylsilane, is used as an intermediates of OLED.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents.