Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetraphenylphthalic anhydride, 98%

$84.97 - $295.54

Chemical Identifiers

Chemical Name or Material Tetraphenylphthalic anhydride
CAS 4741-53-1
Molecular Formula C32H20O3
Formula Weight 452.51
MDL Number MFCD00044504
Beilstein 353504
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1199806

  SDS
alfa aesar™
A1199806
5g Each for $84.97

AAA1199809

  SDS
alfa aesar™
A1199809
10g Each for $148.85

AAA1199814

  SDS
alfa aesar™
A1199814
25g Each for $295.54
Description & Specifications

Specifications

Formula Weight 452.51
Chemical Name or Material Tetraphenylphthalic anhydride
TSCA TSCA
Assay 98%
Molecular Formula C32H20O3
MDL Number MFCD00044504
CAS 4741-53-1
Beilstein 353504
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 289° to 297°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.