missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetraphenylphosphonium tetrafluoroborate, 98+%

Manufacturer:  Alfa Aesar™ A1514130

 View more versions of this product

Catalog No. AAA1514130



Specifications

Specifications

Tetraphenylphosphonium tetrafluoroborate
>300°C
426-79-9
(C6H5)4PBF4
3834618
MAJBFAYVSDYMQG-UHFFFAOYSA-N
tetraphenylphosphanium;tetrafluoroborate
2734204
≥98%
Odorless
250g
C24H20BF4P
MFCD00051889
tetraphenylphosphonium tetrafluoroborate, tetraphenylphosphanium tetrafluoroborate, acmc-20ajix, tetraphenyl-phosphonium fluoborate
[B-](F)(F)(F)F.C1=CC=C(C=C1)[P+](C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4
426.201
426.2

Chemical Identifiers

426-79-9
426.201
MAJBFAYVSDYMQG-UHFFFAOYSA-N
2734204
[B-](F)(F)(F)F.C1=CC=C(C=C1)[P+](C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4
C24H20BF4P
MFCD00051889
tetraphenylphosphonium tetrafluoroborate, tetraphenylphosphanium tetrafluoroborate, acmc-20ajix, tetraphenyl-phosphonium fluoborate
tetraphenylphosphanium;tetrafluoroborate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

Documents