Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetraphenylphosphonium tetrafluoroborate, 98+%

$1,490.19 - $1,490.19

Chemical Identifiers

Chemical Name or Material Tetraphenylphosphonium tetrafluoroborate
CAS 426-79-9
Molecular Formula C24H20BF4P
Linear Formula (C6H5)4PBF4
Formula Weight 426.2
MDL Number MFCD00051889
Beilstein 3834618
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1514130

  SDS
alfa aesar™
A1514130
250g Each for $1,490.19
Description & Specifications

Specifications

Formula Weight 426.2
Molecular Formula C24H20BF4P
Chemical Name or Material Tetraphenylphosphonium tetrafluoroborate
MDL Number MFCD00051889
Melting Point >300°C
Odor Odorless
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Linear Formula (C6H5)4PBF4
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
CAS 426-79-9
Quantity 250g
Assay ≥98%
Beilstein 3834618
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.