Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetraphenylphosphonium hexafluoroantimonate, 98+%

$108.07 - $108.07

Chemical Identifiers

Chemical Name or Material Tetraphenylphosphonium hexafluoroantimonate
CAS 124329-50-6
Molecular Formula (C5H5)4PSbF6
Formula Weight 575.13
MDL Number MFCD00051888
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2475806

  SDS
alfa aesar™
B2475806
5g Each for $108.07
Description & Specifications

Specifications

Formula Weight 575.13
Chemical Name or Material Tetraphenylphosphonium hexafluoroantimonate
Assay ≥98%
Molecular Formula (C5H5)4PSbF6
MDL Number MFCD00051888
CAS 124329-50-6
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P261-P264-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P261-P264-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.