missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetraphenylphosphonium chloride, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 204030100

 View more versions of this product

Catalog No. AC204030100



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for C-C Bond Formation (Olefination)
Specifications

Specifications

Tetraphenylphosphonium chloride
2001-45-8
White to Beige
97.5% min. (Argentometry)
C24H20ClP
MFCD00011916
16, II, 373
Solubility in water: soluble.
[Cl-].C1=CC=C(C=C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
374.85
374.84
Crystalline Powder
99%
275.0°C to 280.0°C
Authentic
Glass bottle
(C6H5)4PCl
10 g
tetraphenylphosphonium chloride, phosphonium, tetraphenyl-, chloride, tetraphenylphosphoniumchloride, tetraphenylchlorophosphine, tetraphenylphosphanium chloride, tetraphenyl phosphonium chloride, ph4pcl, tetraphenylphophonium chloride, tetraphenylphosphonium chloride, for the spectrophotometric det. of bi, co
WAGFXJQAIZNSEQ-UHFFFAOYSA-M
tetraphenylphosphanium;chloride
164911
98%

Chemical Identifiers

2001-45-8
374.85
WAGFXJQAIZNSEQ-UHFFFAOYSA-M
164911
[Cl-].C1=CC=C(C=C1)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
C24H20ClP
MFCD00011916
tetraphenylphosphonium chloride, phosphonium, tetraphenyl-, chloride, tetraphenylphosphoniumchloride, tetraphenylchlorophosphine, tetraphenylphosphanium chloride, tetraphenyl phosphonium chloride, ph4pcl, tetraphenylphophonium chloride, tetraphenylphosphonium chloride, for the spectrophotometric det. of bi, co
tetraphenylphosphanium;chloride
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee

GHS Signal Word: Warning

EINECSNumber : 217-890-3

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only