Learn More
Tetraphenylphosphine imide, 97%, Thermo Scientific Chemicals
$74.63 - $221.58
Chemical Identifiers
CAS | 2325-27-1 |
---|---|
Molecular Formula | C24H20NP |
Molecular Weight (g/mol) | 353.41 |
MDL Number | MFCD00003044 |
InChI Key | PTLOPIHJOPWUNN-UHFFFAOYSA-N |
Synonym | tetraphenylphosphine imide, n-triphenylphosphoranylidene aniline, phenylimino triphenylphosphorane, phosphine imide, tetraphenyl, benzenamine, n-triphenylphosphoranylidene, tetraphenylphosphinimine, triphenylphosphine phenylimide, benzenamine,n-triphenylphosphoranylidene, n,1,1,1-tetraphenyl-??-phosphanimine, triphenyl phenylimino |
PubChem CID | 75352 |
IUPAC Name | triphenyl(phenylimino)-$l^{5}-phosphane |
SMILES | C1=CC=C(C=C1)N=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL0049603
|
Thermo Scientific Chemicals
L0049603 |
1 g |
N/A
|
|
|||||
AAL0049606
|
Thermo Scientific Chemicals
L0049606 |
5 g |
N/A
|
|
|||||
Description
Has been used as a protected form of aniline for the preparation of monoalkylanilines by alkylation and hydrolysis and as an intermediate for the preparation of isocyanates and isothiocyanates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsHas been used as a protected form of aniline for the preparation of monoalkylanilines by alkylation and hydrolysis and as an intermediate for the preparation of isocyanates and isothiocyanates.
Solubility
Insoluble in water. Soluble in chloroform 25 mg/mL.
Notes
Moisture sensitive. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
Chemical Identifiers
2325-27-1 | |
353.41 | |
PTLOPIHJOPWUNN-UHFFFAOYSA-N | |
75352 | |
C1=CC=C(C=C1)N=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
C24H20NP | |
MFCD00003044 | |
tetraphenylphosphine imide, n-triphenylphosphoranylidene aniline, phenylimino triphenylphosphorane, phosphine imide, tetraphenyl, benzenamine, n-triphenylphosphoranylidene, tetraphenylphosphinimine, triphenylphosphine phenylimide, benzenamine,n-triphenylphosphoranylidene, n,1,1,1-tetraphenyl-??-phosphanimine, triphenyl phenylimino | |
triphenyl(phenylimino)-$l^{5}-phosphane |
Specifications
2325-27-1 | |
Odorless | |
(C6H5)3P=N(C6H5) | |
1 g | |
Moisture sensitive | |
Insoluble in water. Soluble in chloroform 25mg/mL. | |
C1=CC=C(C=C1)N=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 | |
353.41 | |
353.4 | |
Tetraphenylphosphine imide |
128°C to 132°C | |
C24H20NP | |
MFCD00003044 | |
753032 | |
tetraphenylphosphine imide, n-triphenylphosphoranylidene aniline, phenylimino triphenylphosphorane, phosphine imide, tetraphenyl, benzenamine, n-triphenylphosphoranylidene, tetraphenylphosphinimine, triphenylphosphine phenylimide, benzenamine,n-triphenylphosphoranylidene, n,1,1,1-tetraphenyl-??-phosphanimine, triphenyl phenylimino | |
PTLOPIHJOPWUNN-UHFFFAOYSA-N | |
triphenyl(phenylimino)-$l^{5}-phosphane | |
75352 | |
97% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 219-039-1
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only