missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetraphenylphosphine imide, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L0049606

 View more versions of this product

Catalog No. AAL0049606



Description

Description

Has been used as a protected form of aniline for the preparation of monoalkylanilines by alkylation and hydrolysis and as an intermediate for the preparation of isocyanates and isothiocyanates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Has been used as a protected form of aniline for the preparation of monoalkylanilines by alkylation and hydrolysis and as an intermediate for the preparation of isocyanates and isothiocyanates.

Solubility
Insoluble in water. Soluble in chloroform 25 mg/mL.

Notes
Moisture sensitive. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
Specifications

Specifications

Tetraphenylphosphine imide
128°C to 132°C
C24H20NP
MFCD00003044
753032
tetraphenylphosphine imide, n-triphenylphosphoranylidene aniline, phenylimino triphenylphosphorane, phosphine imide, tetraphenyl, benzenamine, n-triphenylphosphoranylidene, tetraphenylphosphinimine, triphenylphosphine phenylimide, benzenamine,n-triphenylphosphoranylidene, n,1,1,1-tetraphenyl-??-phosphanimine, triphenyl phenylimino
PTLOPIHJOPWUNN-UHFFFAOYSA-N
triphenyl(phenylimino)-$l^{5}-phosphane
75352
97%
2325-27-1
Odorless
(C6H5)3P=N(C6H5)
5 g
Moisture sensitive
Insoluble in water. Soluble in chloroform 25mg/mL.
C1=CC=C(C=C1)N=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
353.41
353.4

Chemical Identifiers

2325-27-1
353.41
PTLOPIHJOPWUNN-UHFFFAOYSA-N
75352
C1=CC=C(C=C1)N=P(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
C24H20NP
MFCD00003044
tetraphenylphosphine imide, n-triphenylphosphoranylidene aniline, phenylimino triphenylphosphorane, phosphine imide, tetraphenyl, benzenamine, n-triphenylphosphoranylidene, tetraphenylphosphinimine, triphenylphosphine phenylimide, benzenamine,n-triphenylphosphoranylidene, n,1,1,1-tetraphenyl-??-phosphanimine, triphenyl phenylimino
triphenyl(phenylimino)-$l^{5}-phosphane
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 219-039-1

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only