Learn More
Tetraphenylboron sodium, 99.5%, Thermo Scientific Chemicals
Tetraphenylboron sodium, 99.5%, C24H20BNa, CAS Number-143-66-8, borate 1-, tetraphenyl-, sodium, dotite kalibor, kalibor, kalignost, kariporu k, sodium tetraphenylborate, sodium tetraphenylborate 1-, sodium tetraphenylboride, sodium tetraphenylboron, tetraphenylboron sodium, 100g
Supplier: Thermo Scientific Chemicals 138520250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Tetraphenylboron sodium | |
143-66-8 | |
Colorless | |
Authentic | |
Glass bottle | |
(C6H5)4BNa | |
25 g | |
sodium tetraphenylborate, tetraphenylboron sodium, sodium tetraphenylboron, kalignost, sodium tetraphenylborate 1-, kalibor, dotite kalibor, kariporu k, borate 1-, tetraphenyl-, sodium, sodium tetraphenylboride | |
HFSRCEJMTLMDLI-UHFFFAOYSA-N | |
sodium;tetraphenylboranuide | |
2723787 | |
99.5% |
p.a. | |
>300.0°C | |
0.5 % max. (105°C) | |
99.4 % min. (Total base) | |
C24H20BNa | |
MFCD00011494 | |
15, 8818 | |
Solubility in water: 50g/L (20°C). Other solubilities: freely soluble in acetone, slightly soluble in ether and chloroform, practically insoluble in petroleum ether | |
[Na+].C1=CC=C(C=C1)[B-](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 | |
342.22 | |
342.21 | |
Liquid |
Chemical Identifiers
143-66-8 | |
342.22 | |
HFSRCEJMTLMDLI-UHFFFAOYSA-N | |
2723787 | |
[Na+].C1=CC=C(C=C1)[B-](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
C24H20BNa | |
MFCD00011494 | |
sodium tetraphenylborate, tetraphenylboron sodium, sodium tetraphenylboron, kalignost, sodium tetraphenylborate 1-, kalibor, dotite kalibor, kariporu k, borate 1-, tetraphenyl-, sodium, sodium tetraphenylboride | |
sodium;tetraphenylboranuide |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
GHS P Statement
Call a POISON CENTER or doctor/physician if you feel unwell.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective cloth
GHS Signal Word: Danger
EINECSNumber : 205-605-5
RTECSNumber : ED3362500
TSCA : TSCA
RUO – Research Use Only