Please login to your online account to display your discounted pricing

Tetraphenylboron sodium, 99.5%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  138521000

 View more versions of this product

Catalog No. AC138521000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99.4% (Total base)
Physical Form Liquid
Formula Weight 342.21
Quantity 100g
Color Clear colorless
Health Hazard 3 GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing.
EINECS Number 205-605-5
TSCA TSCA
RTECS Number ED3362500
Merck Index 15, 8818
Chemical Name or Material Tetraphenylboron sodium
Infrared Spectrum Authentic
Loss on Drying 0.5 % max. (105°C)
Packaging Glass bottle
Name Note p.a.
Molecular Formula C24H20BNa
Linear Formula (C6H5)4BNa
Synonym Sodium tetraphenylborate
Health Hazard 1 Danger
Health Hazard 2 GHS P Statement
Toxic if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Assay Percent Range 99.5%
MDL Number MFCD00011494
CAS 143-66-8

Specifications

Chemical Name or Material Tetraphenylboron sodium
Name Note p.a.
CAS 143-66-8
Assay ≥99.4% (Total base)
Assay Percent Range 99.5%
Molecular Formula C24H20BNa
Linear Formula (C6H5)4BNa
Formula Weight 342.21
MDL Number MFCD00011494
Synonym Sodium tetraphenylborate
Accessories

Danger

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing.

GHS P Statement
Toxic if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

SDS & Safety Documents