missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetraphenylarsonium chloride monohydrate, 97%, ACROS Organics™

Chemical Identifiers

CAS 104170-16-3
Molecular Formula C24H22AsClO
Molecular Weight (g/mol) 436.811
MDL Number MFCD00149995
InChI Key NGDWSDTZKCSLRK-UHFFFAOYSA-M
Synonym acmc-20m6yd
PubChem CID 2723786
IUPAC Name tetraphenylarsanium;chloride;hydrate
SMILES C1=CC=C(C=C1)[As+](C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4.O.[Cl-]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC366950100 View Documents Thermo Fisher Scientific
AC366950100
10g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Phase transfer catalyst
Specifications

Chemical Identifiers

104170-16-3
436.811
NGDWSDTZKCSLRK-UHFFFAOYSA-M
2723786
C1=CC=C(C=C1)[As+](C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4.O.[Cl-]
C24H22AsClO
MFCD00149995
acmc-20m6yd
tetraphenylarsanium;chloride;hydrate

Specifications

Authentic
White to Beige
104170-16-3
C24H22AsClO
MFCD00149995
NGDWSDTZKCSLRK-UHFFFAOYSA-M
tetraphenylarsanium;chloride;hydrate
2723786
Crystalline Powder
Tetraphenylarsonium chloride monohydrate, 97%
Glass bottle
10g
97%
(C6H5)4AsCl·H2O
acmc-20m6yd
C1=CC=C(C=C1)[As+](C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4.O.[Cl-]
436.811
436.82
≥96.0%
Safety and Handling

Safety and Handling

GHS H Statement:
Very toxic to aquatic life with long lasting effects.
Toxic if swallowed.
Toxic if inhaled.

GHS P Statement:
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger