Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetraphenylarsonium chloride hydrate, 96%

Chemical Identifiers

Chemical Name or Material Tetraphenylarsonium chloride hydrate
CAS 507-28-8
Molecular Formula (C6H5)4AsCl·xH2O
Formula Weight 418.80g/mol (Anhydrous)
MDL Number MFCD00011908
UN Number UN3465
Beilstein 3580063
Merck Index 14,9236
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1604303

 
alfa aesar™
A16043-03
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 256° to 259°C
Quantity 1g
Chemical Name or Material Tetraphenylarsonium chloride hydrate
CAS 507-28-8
Assay 96%
Molecular Formula (C6H5)4AsCl·xH2O
Formula Weight 418.80g/mol (Anhydrous)
MDL Number MFCD00011908
UN Number UN3465
Beilstein 3580063
Merck Index 14,9236
Solubility Information Freely soluble in water; Soluble in alcohol; Sparingly soluble in acetone
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.
Health Hazard 3 GHS P Statement
P261-P301+P310-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOARSENIC COMPOUND, SOLID, N.O.S.

Applications
Tetraphenylarsonium chloride hydrate is a cation exchange reagent. It is used to prepare arsonium amide, oxime, and carbohydrate derivatives.

Solubility
Freely soluble in water. Soluble in alcohol. Sparingly soluble in acetone

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P261-P301+P310-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.