Please login to your online account to display your discounted pricing

Tetrapentylammonium bromide, 99+%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  221480050

 View more versions of this product

Catalog No. AC221480050

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99.0%
Physical Form Shiny Flakes
Formula Weight 378.47
Quantity 5g
Color White
Beilstein 04, III, 331
Melting Point 100° to 102°C
Chemical Name or Material Tetrapentylammonium bromide
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Infrared Spectrum Authentic
Packaging Glass bottle
Assay Percent Range 99+%
Molecular Formula C20H44BrN
Linear Formula {CH3(CH2)4}4NBr
MDL Number MFCD00011856
CAS 866-97-7
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Tetrapentylammonium bromide
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C20H44BrN
Linear Formula {CH3(CH2)4}4NBr
Formula Weight 378.47
MDL Number MFCD00011856
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

SDS & Safety Documents