Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetraoctadecylammonium bromide, 98%

Chemical Identifiers

Chemical Name or Material Tetraoctadecylammonium bromide
CAS 63462-99-7
Formula Weight 1107.9
MDL Number MFCD00043172
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA2058206

 
alfa aesar™
2058206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 1107.9
Chemical Name or Material Tetraoctadecylammonium bromide
Physical Form Crystalline
CAS 63462-99-7
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 98%
MDL Number MFCD00043172
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Sensitivity Hygroscopic
Melting Point 103° to 104°C
Quantity 5g