Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetramethyltin, 98%

Chemical Identifiers

Chemical Name or Material Tetramethyltin
CAS 594-27-4
Molecular Formula (CH3)4Sn
Formula Weight 178.83
MDL Number MFCD00008278
UN Number UN3384
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA7114504

  SDS
alfa aesar™
7114504
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA7114509

  SDS
alfa aesar™
7114509
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA7114518

  SDS
alfa aesar™
7114518
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 178.83
Physical Form Liquid
Solubility Information Insoluble in water; Soluble in organic solvents
Molecular Formula (CH3)4Sn
Quantity 2g
Health Hazard 1 Danger
Assay 98%
MDL Number MFCD00008278
Health Hazard 2 GHS H Statement
H225-H300-H310-H330
Highly flammable liquid and vapor.
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Refractive Index 1.4410
TSCA TSCA
Flash Point −12°C (9°F)
Chemical Name or Material Tetramethyltin
Odor Unpleasant
Density 1.291g/mL
RTECS Number WH8630000
Boiling Point 74° to 75°C
DOT Information Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TOXIC BY INHALATION LIQUID, FLAMMABLE, N.O.S.
UN Number UN3384
CAS 594-27-4
Melting Point −54°C
Health Hazard 3 GHS P Statement
P210-P301+P310-P303+P361+P353-P304+P340-P320-P330-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P210-P301+P310-P303+P361+P353-P304+P340-P320-P330-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H300-H310-H330
Highly flammable liquid and vapor.
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.