missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetramethyltin, 98%

Manufacturer:  Alfa Aesar™ 7114509

 View more versions of this product

Catalog No. AA7114509



Specifications

Specifications

Tetramethyltin
-54°C
10g
Unpleasant
1.4410
MFCD00008278
tetramethyltin, stannane, tetramethyl, tetramethyl tin, tin, tetramethyl, tetramethylcin, snme4, ch3 4sn, tetramethylcin czech, tin tetramethyl, unii-8v4xu9dpbk
VXKWYPOMXBVZSJ-UHFFFAOYSA-N
tetramethylstannane
11661
178.83
98%
74°C to 75°C
594-27-4
−12°C (9°F)
1.291g/mL
C4H12Sn
UN3384
Insoluble in water; Soluble in organic solvents
C[Sn](C)(C)C
178.85
CHEBI:30420
Liquid

Chemical Identifiers

594-27-4
178.85
VXKWYPOMXBVZSJ-UHFFFAOYSA-N
11661
tetramethylstannane
C4H12Sn
MFCD00008278
tetramethyltin, stannane, tetramethyl, tetramethyl tin, tin, tetramethyl, tetramethylcin, snme4, ch3 4sn, tetramethylcin czech, tin tetramethyl, unii-8v4xu9dpbk
CHEBI:30420
C[Sn](C)(C)C
Safety and Handling

Safety and Handling

GHS H Statement
H225-H300-H310-H330
Highly flammable liquid and vapor.
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.

GHS P Statement
P210-P301+P310-P303+P361+P353-P304+P340-P320-P330-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TOXIC BY INHALATION LIQUID, FLAMMABLE, N.O.S.

RTECSNumber : WH8630000

TSCA : TSCA

SDS Documents