Please login to your online account to display your discounted pricing

Tetramethylthiuram Disulfide 97%, ACROS Organics™

$23.32 - $104.18

Chemical Identifiers

Chemical Name or Material Tetramethylthiuram disulfide
Name Note 97%
CAS 137-26-8
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C6H12N2S4
Linear Formula [(CH3)2NCS2]2
Formula Weight 240.41
MDL Number MFCD00008325
Synonym Bis(dimethylthiocarbamyl) disulfide
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC138480010

  SDS
thermo fisher scientific
138480010
Plastic bottle 1kg Each for $104.18

AC138480050

  SDS
thermo fisher scientific
138480050
Glass bottle 5g Each for $23.32

AC138485000

  SDS
thermo fisher scientific
138485000
Plastic bottle 500g Each for $69.03
Description & Specifications

Specifications

Assay ≥96.0%
Formula Weight 240.41
Physical Form Powder
Melting Point >142°C
Quantity 1kg
Color White to off-white
Beilstein 04, 76
Merck Index 15, 9525
TSCA TSCA
EINECS Number 205-286-2
RTECS Number JO1400000
Synonym Bis(dimethylthiocarbamyl) disulfide
Ash 0.5% max.
Health Hazard 3 GHS P Statement:
Wear eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 GHS Signal Word: Warning
Particle Size residue on 150 micron sieve: 0.1% max.
Additive 0.5 to 2.5% oil
Infrared Spectrum Authentic
Loss on Drying 0.5% max.
Packaging Plastic bottle
Health Hazard 2 GHS H Statement:
May cause an allergic skin reaction.
Harmful if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Very toxic to aquatic life with long lasting effects.
Causes serious eye irritation.
Harmful if swallowed.
Linear Formula [(CH3)2NCS2]2
Molecular Formula C6H12N2S4
MDL Number MFCD00008325
Chemical Name or Material Tetramethylthiuram disulfide
CAS 137-26-8
Name Note 97%
Assay Percent Range 97%
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Wear eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause an allergic skin reaction.
Harmful if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Very toxic to aquatic life with long lasting effects.
Causes serious eye irritation.
Harmful if swallowed.