missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetramethyltetraselenafulvalene, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 346500500

 View more versions of this product

Catalog No. AC346500500

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

448.04
50mg
Glass bottle
C10H12Se4
Tetramethyltetraselenafulvalene
97%
Crystalline Powder
Red-violet
Complies, 96.0% min.
55259-49-9
97%
MFCD00011507

Chemical Identifiers

55259-49-9
C10H12Se4
MFCD00011507
97%
448.04
Tetramethyltetraselenafulvalene, 97%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
May cause damage to organs through prolonged or repeated exposure.
Toxic if swallowed.
Toxic if inhaled.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger