Learn More
Tetramethylethylene, 97%, Thermo Scientific Chemicals
$69.10 - $731.29
Chemical Identifiers
CAS | 563-79-1 |
---|---|
Molecular Formula | C6H12 |
Molecular Weight (g/mol) | 84.162 |
MDL Number | MFCD00008897 |
InChI Key | WGLLSSPDPJPLOR-UHFFFAOYSA-N |
Synonym | 2,3-dimethyl-2-butene, tetramethylethylene, 2-butene, 2,3-dimethyl, 2,3-dimethylbutene-2, 1,1,2,2-tetramethylethylene, tetramethylethene, 2,3-dimethyl-but-2-ene, ch3 2c=c ch3 2, 2-butene,3-dimethyl, acmc-1akps |
PubChem CID | 11250 |
IUPAC Name | 2,3-dimethylbut-2-ene |
SMILES | CC(=C(C)C)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1320106
|
Thermo Scientific Chemicals
A1320106 |
5 g |
Each for $69.10
|
|
|||||
AAA1320114
|
Thermo Scientific Chemicals
A1320114 |
25 g |
Each for $207.99
|
|
|||||
AAA1320122
|
Thermo Scientific Chemicals
A1320122 |
100 g |
Each for $731.29
|
|
|||||
Description
Tetramethylethylene is an pharmaceutical intermediate. Ozonolysys in neat tetramethyleneethylene gives tetramethyleneethylene epoxide as product.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTetramethylethylene is an pharmaceutical intermediate. Ozonolysys in neat tetramethyleneethylene gives tetramethyleneethylene epoxide as product.
Solubility
Soluble in water (0.071 g/L)
Notes
Store at room temperature. Incompatible with oxidizing agents. Air sensitive.
Chemical Identifiers
563-79-1 | |
84.162 | |
WGLLSSPDPJPLOR-UHFFFAOYSA-N | |
11250 | |
CC(=C(C)C)C |
C6H12 | |
MFCD00008897 | |
2,3-dimethyl-2-butene, tetramethylethylene, 2-butene, 2,3-dimethyl, 2,3-dimethylbutene-2, 1,1,2,2-tetramethylethylene, tetramethylethene, 2,3-dimethyl-but-2-ene, ch3 2c=c ch3 2, 2-butene,3-dimethyl, acmc-1akps | |
2,3-dimethylbut-2-ene |
Specifications
563-79-1 | |
0.705 | |
−17°C (1°F) | |
C6H12 | |
MFCD00008897 | |
UN3295 | |
Air Sensitive | |
Soluble in water (0.071g/L) | |
CC(=C(C)C)C | |
84.162 | |
84.16 | |
Tetramethylethylene |
-75°C | |
72°C to 73°C | |
Mild | |
1.412 | |
5 g | |
1361357 | |
2,3-dimethyl-2-butene, tetramethylethylene, 2-butene, 2,3-dimethyl, 2,3-dimethylbutene-2, 1,1,2,2-tetramethylethylene, tetramethylethene, 2,3-dimethyl-but-2-ene, ch3 2c=c ch3 2, 2-butene,3-dimethyl, acmc-1akps | |
WGLLSSPDPJPLOR-UHFFFAOYSA-N | |
2,3-dimethylbut-2-ene | |
11250 | |
97% |
Safety and Handling
GHS H Statement
H225-H304
Highly flammable liquid and vapour.
May be fatal if swallowed and enters airways.
P210-P233-P240-P241-P242-P243-P280-P301+P310-P303+P361+P353-P331-P372-P374-P380-P501c
H225-H304
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
EINECSNumber : 209-263-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only