Please login to your online account to display your discounted pricing

Tetramethylene Sulfoxide, 98+%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: fisher scientific acros  138440250

 View more versions of this product

Catalog No. AC138440250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Assay ≥98.0%
Formula Weight 104.17
Quantity 25g
Color Clear colorless to yellow
Fieser 01,1145
TSCA TSCA
RTECS Number XN0830000
Density 1.1500g/mL
EINECS Number 216-493-2
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.5180 to 1.5220 (20°C, 589nm)
Synonym Tetrahydrothiophene oxide
CAS 1600-44-8
Molecular Formula C4H8OS
Name Note 98+%
Chemical Name or Material Tetramethylene sulfoxide
MDL Number MFCD00005477
Assay Percent Range 98+%

Specifications

Chemical Name or Material Tetramethylene sulfoxide
Name Note 98+%
CAS 1600-44-8
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C4H8OS
Formula Weight 104.17
MDL Number MFCD00005477
Synonym Tetrahydrothiophene oxide