missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Tetramethylene sulfone, 99+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 138430025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Tetramethylene sulfone | |
27.4°C to 27.8°C | |
1.2600g/mL | |
165°C | |
99% min. (GC) | |
C4H8O2S | |
MFCD00005484 | |
17,I,5 | |
1.26 | |
sulfolane, tetramethylene sulfone, tetrahydrothiophene 1,1-dioxide, sulfolan, sulpholane, sulfalone, sulphoxaline, 1,1-dioxothiolan, dioxothiolan, thiophan sulfone | |
HXJUTPCZVOIRIF-UHFFFAOYSA-N | |
thiolane 1,1-dioxide | |
31347 | |
CHEBI:74794 | |
99+% |
126-33-0 | |
Yellow | |
285.0°C | |
Authentic | |
Glass bottle | |
1.4820 to 1.4850 | |
2.5 L | |
01,1144; 02,402; 05,651; 13,256 | |
15,9085 | |
Solubility in water: soluble. Other solubilities: soluble in diluted mineral acids, soluble in acetone, benzene and toluene, freely soluble in alcohol | |
O=S1(=O)CCCC1 | |
120.17 | |
10.34 mPa.s (30°C) | |
120.17 | |
Liquid After Melting |
Chemical Identifiers
126-33-0 | |
120.17 | |
HXJUTPCZVOIRIF-UHFFFAOYSA-N | |
31347 | |
thiolane 1,1-dioxide |
C4H8O2S | |
MFCD00005484 | |
sulfolane, tetramethylene sulfone, tetrahydrothiophene 1,1-dioxide, sulfolan, sulpholane, sulfalone, sulphoxaline, 1,1-dioxothiolan, dioxothiolan, thiophan sulfone | |
CHEBI:74794 | |
O=S1(=O)CCCC1 |
Safety and Handling
GHS Signal Word: Warning
EINECSNumber : 204-783-1
RUO – Research Use Only