Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetramethylcyclopentadiene, mixed isomers, tech. 85%

Chemical Identifiers

Chemical Name or Material Tetramethylcyclopentadiene
CAS 4249-10-9
Name Note Mixed isomers
Purity Grade Technical Grade
Formula Weight 122.21
MDL Number MFCD00145385
UN Number UN3295
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3857703

  SDS
alfa aesar™
3857703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA3857706

  SDS
alfa aesar™
3857706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 122.21
MDL Number MFCD00145385
Flash Point 42° (108°F)
Health Hazard 2 GHS H Statement
H226
Flammable liquid and vapor.
Name Note Mixed isomers
CAS 4249-10-9
UN Number UN3295
Boiling Point 55° to 59°C (13mmHg)
Refractive Index 1.4720
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Physical Form Liquid
Quantity 1g
Sensitivity Air & Moisture Sensitive
Health Hazard 1 Warning
Color Colorless
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
Chemical Name or Material Tetramethylcyclopentadiene
Purity Grade Technical Grade
Density 0.808g/mL
Assay 85%
Safety and Handling

Warning

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226
Flammable liquid and vapor.