Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetramethylammonium tribromide, 98%

Chemical Identifiers

Chemical Name or Material Tetramethylammonium tribromide
CAS 15625-56-6
Molecular Formula (CH3)4NBr3
Formula Weight 313.86
MDL Number MFCD00032165
UN Number UN3261
Beilstein 3914655
Synonym Tetramethylammonium bromide perbromide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2423722

  SDS
alfa aesar™
B2423722
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 313.86
Chemical Name or Material Tetramethylammonium tribromide
TSCA TSCA
Assay 98%
Molecular Formula (CH3)4NBr3
MDL Number MFCD00032165
CAS 15625-56-6
UN Number UN3261
Beilstein 3914655
Synonym Tetramethylammonium bromide perbromide
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 114° to 120°C
Quantity 100g
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.