missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetramethylammonium Triacetoxyborohydride, TCI America™

Manufacturer:  TCI America T155325G

 View more versions of this product

Catalog No. T155325G



Specifications

Specifications

Tetramethylammonium Triacetoxyborohydride
25g
C10H21BNO6
2813
ZWSJBUUORJFGHK-UHFFFAOYSA-N
tetramethylazanium;triacetyloxyboron(1-)
10858300
Crystal-Powder at 20°C
White
109704-53-2
MFCD00012196
tetramethylammonium triacetoxyborohydride, me4nhb oac 3, me4n aco 3bh;, me4nb oac 3h;, me4n bh oac 3;, ch3 4nhb oac ;, me4n ch3co2 3bh;, tetramethylammonium triacetoxyhydroborate, tetramethylammonium triacetoxyborohydride;, bis acetyloxy boranuidyl acetate; tetramethylazanium
[B-](OC(=O)C)(OC(=O)C)OC(=O)C.C[N+](C)(C)C
262.089
263.10
≥85.0% (T)

Chemical Identifiers

109704-53-2
262.089
ZWSJBUUORJFGHK-UHFFFAOYSA-N
10858300
[B-](OC(=O)C)(OC(=O)C)OC(=O)C.C[N+](C)(C)C
C10H21BNO6
MFCD00012196
tetramethylammonium triacetoxyborohydride, me4nhb oac 3, me4n aco 3bh;, me4nb oac 3h;, me4n bh oac 3;, ch3 4nhb oac ;, me4n ch3co2 3bh;, tetramethylammonium triacetoxyhydroborate, tetramethylammonium triacetoxyborohydride;, bis acetyloxy boranuidyl acetate; tetramethylazanium
tetramethylazanium;triacetyloxyboron(1-)
Safety and Handling

Safety and Handling

TSCA : Yes

Documents