missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetramethylammonium perchlorate

Manufacturer:  Alfa Aesar™ 3083414

 View more versions of this product

Catalog No. AA3083414



Specifications

Specifications

Tetramethylammonium perchlorate
25g
C4H12ClNO4
UN1479
ZCWKIFAQRXNZCH-UHFFFAOYSA-M
tetramethylazanium;perchlorate
17337
Crystalline
>300°C
2537-36-2
MFCD00043207
tetramethylammonium perchlorate, methanaminium, n,n,n-trimethyl-, perchlorate, n,n,n-trimethylmethanaminium perchlorate, ammonium, tetramethyl-, perchlorate, methanaminium, n,n,n-trimethyl-, perchlorate 1:1, c4h12n.clo4, acmc-20ajj6, tetramethylazanium perchlorate, tetramethylammonium perchlorate 25g, wln: 2k2&2&2 &wgw
C[N+](C)(C)C.[O-]Cl(=O)(=O)=O
173.593
173.6

Chemical Identifiers

2537-36-2
173.593
ZCWKIFAQRXNZCH-UHFFFAOYSA-M
17337
C[N+](C)(C)C.[O-]Cl(=O)(=O)=O
C4H12ClNO4
MFCD00043207
tetramethylammonium perchlorate, methanaminium, n,n,n-trimethyl-, perchlorate, n,n,n-trimethylmethanaminium perchlorate, ammonium, tetramethyl-, perchlorate, methanaminium, n,n,n-trimethyl-, perchlorate 1:1, c4h12n.clo4, acmc-20ajj6, tetramethylazanium perchlorate, tetramethylammonium perchlorate 25g, wln: 2k2&2&2 &wgw
tetramethylazanium;perchlorate
Safety and Handling

Safety and Handling

GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P221-P210-P220-P305+P351+P338-P405-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Keep/Store away from clothing/combustible materials.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, N.O.S.

TSCA : TSCA

SDS Documents