missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetramethylammonium iodide, 99%, ACROS Organics™

Chemical Identifiers

Assay Percent Range 99%
Molecular Formula C4H12IN
Linear Formula (CH3)4NI
Formula Weight 201.05
MDL Number MFCD00011629
Chemical Name or Material Tetramethylammonium iodide
Assay ≥98.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC216681000 SDS Fisher Scientific Acros
216681000
Glass bottle 100g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

Phase transfer catalyst
Description & Specifications

Chemical Identifiers

99%
(CH3)4NI
MFCD00011629
≥98.5%
C4H12IN
201.05
Tetramethylammonium iodide

Specifications

White
75-58-1
Glass bottle
99%
(CH3)4NI
MFCD00011629
15, 9372
≥98.5%
100g
Authentic
Fine Crystalline Powder
C4H12IN
201.05
04, 51
Tetramethylammonium iodide