Please login to your online account to display your discounted pricing

Tetramethylammonium iodide, 99%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: fisher scientific acros  216681000

 View more versions of this product

Catalog No. AC216681000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Fine Crystalline Powder
Formula Weight 201.05
Quantity 100g
Color White
Beilstein 04, 51
Merck Index 15, 9372
Chemical Name or Material Tetramethylammonium iodide
Infrared Spectrum Authentic
Packaging Glass bottle
Assay Percent Range 99%
Molecular Formula C4H12IN
Linear Formula (CH3)4NI
MDL Number MFCD00011629
CAS 75-58-1
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Tetramethylammonium iodide
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C4H12IN
Linear Formula (CH3)4NI
Formula Weight 201.05
MDL Number MFCD00011629
Accessories

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents