Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetramethylammonium iodide, 99%

Chemical Identifiers

Chemical Name or Material Tetramethylammonium iodide
CAS 75-58-1
Molecular Formula C4H12IN
Linear Formula (CH3)4NI
Formula Weight 201.05
MDL Number MFCD00011629
UN Number UN2811
Beilstein 3620030
Merck Index 14,9225
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1281122

 
alfa aesar™
A1281122
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 201.05
Chemical Name or Material Tetramethylammonium iodide
TSCA TSCA
Assay 99%
Molecular Formula C4H12IN
Linear Formula (CH3)4NI
MDL Number MFCD00011629
CAS 75-58-1
UN Number UN2811
Beilstein 3620030
Merck Index 14,9225
Solubility Information Slightly soluble in water; Insoluble in chloroform, ether
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280f-P305+P351+P338-P309-P310
Wear protective clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
RTECS Number PA1050000
Odor Odorless
Melting Point >300°C
Quantity 100g
Safety and Handling

Danger

GHS P Statement
P280f-P305+P351+P338-P309-P310
Wear protective clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.