missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetramethylammonium Hydroxide Pentahydrate, TCI America™

Manufacturer:  TCI America T1404100G

 View more versions of this product

Catalog No. T1404100G



Specifications

Specifications

Tetramethylammonium Hydroxide Pentahydrate
73°C
10424-65-4
MFCD00149566
tetramethylammonium hydroxide pentahydrate, b2hkt2lckq, unii-b2hkt2lckq, methanaminium, n,n,n-trimethyl-, hydroxide, pentahydrate, tmaoh, tetramethylammoniumhydroxid, tetrametanoamonio hydroxido, tetrametiloamonio hydroxido, methanaminium, n,n,n-trimethyl-, hydroxide, hydrate 1:1:5, acmc-2098bm
C[N+](C)(C)C.O.O.O.O.O.[OH-]
181.229
91.15
≥97.0% (T)
White
100g
C4H23NO6
3423
MYXKPFMQWULLOH-UHFFFAOYSA-M
tetramethylazanium;hydroxide;pentahydrate
82620
Crystal-Powder at 20°C

Chemical Identifiers

10424-65-4
181.229
MYXKPFMQWULLOH-UHFFFAOYSA-M
82620
C[N+](C)(C)C.O.O.O.O.O.[OH-]
C4H23NO6
MFCD00149566
tetramethylammonium hydroxide pentahydrate, b2hkt2lckq, unii-b2hkt2lckq, methanaminium, n,n,n-trimethyl-, hydroxide, pentahydrate, tmaoh, tetramethylammoniumhydroxid, tetrametanoamonio hydroxido, tetrametiloamonio hydroxido, methanaminium, n,n,n-trimethyl-, hydroxide, hydrate 1:1:5, acmc-2098bm
tetramethylazanium;hydroxide;pentahydrate
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0186

RTECSNumber : PA0875000

TSCA : Yes

Documents