missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetramethylammonium Hydroxide (ca. 25% in Water), TCI America™

Manufacturer:  TCI America T146025ML

 View more versions of this product

Catalog No. T146025ML



Specifications

Specifications

Tetramethylammonium Hydroxide (ca. 25% in Water)
25mL
75-59-2
MFCD00008280
tetramethylammonium hydroxide, tmah, hydroxyde de tetramethylammonium, nmw-w, nmd 3, tetramethyl ammonium hydroxide, unii-5gkp7317q2, ammonium, tetramethyl-, hydroxide, methanaminium, n,n,n-trimethyl-, hydroxide, tetramethylammoniumhydroxide
C[N+](C)(C)C.[OH-]
91.154
91.15
Yellow
>13 (20°C)
C4H13NO
1835
WGTYBPLFGIVFAS-UHFFFAOYSA-M
tetramethylazanium;hydroxide
60966
Clear Liquid at 20°C

Chemical Identifiers

75-59-2
91.154
WGTYBPLFGIVFAS-UHFFFAOYSA-M
60966
C[N+](C)(C)C.[OH-]
C4H13NO
MFCD00008280
tetramethylammonium hydroxide, tmah, hydroxyde de tetramethylammonium, nmw-w, nmd 3, tetramethyl ammonium hydroxide, unii-5gkp7317q2, ammonium, tetramethyl-, hydroxide, methanaminium, n,n,n-trimethyl-, hydroxide, tetramethylammoniumhydroxide
tetramethylazanium;hydroxide
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0186

RTECSNumber : PA0875000

TSCA : Yes

Documents