missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetramethylammonium hydroxide, 25% w/w in methanol

Manufacturer:  Alfa Aesar™ 3083309

 View more versions of this product

Catalog No. AA3083309



Specifications

Specifications

Tetramethylammonium hydroxide
12° (54°F)
Air Sensitive
10g
C4H13NO
MFCD00008280
14,9224
25% w/w in methanol
C[N+](C)(C)C.[OH-]
91.154
91.15
0.865g/mL
Like alcohol
Colorless to Yellow
75-59-2
(CH3)4NOH
UN2924
tetramethylammonium hydroxide, tmah, hydroxyde de tetramethylammonium, nmw-w, nmd 3, tetramethyl ammonium hydroxide, unii-5gkp7317q2, ammonium, tetramethyl-, hydroxide, methanaminium, n,n,n-trimethyl-, hydroxide, tetramethylammoniumhydroxide
WGTYBPLFGIVFAS-UHFFFAOYSA-M
tetramethylazanium;hydroxide
60966
Liquid

Chemical Identifiers

75-59-2
91.154
WGTYBPLFGIVFAS-UHFFFAOYSA-M
60966
C[N+](C)(C)C.[OH-]
C4H13NO
MFCD00008280
tetramethylammonium hydroxide, tmah, hydroxyde de tetramethylammonium, nmw-w, nmd 3, tetramethyl ammonium hydroxide, unii-5gkp7317q2, ammonium, tetramethyl-, hydroxide, methanaminium, n,n,n-trimethyl-, hydroxide, tetramethylammoniumhydroxide
tetramethylazanium;hydroxide
Safety and Handling

Safety and Handling

GHS H Statement
H225-H300-H310-H331-H370-H372-H314
Highly flammable liquid and vapor.
Fatal if swallowed.
Fatal in contact with skin.
Toxic if inhaled.
Causes damage to organs.
Causes damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.

GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.

TSCA : TSCA

SDS Documents