missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetramethylammonium hydroxide, 2.38% w/w aq. soln., Electronic Grade, 99.9999% (metals basis)

Manufacturer:  Alfa Aesar™ 44940K2

 View more versions of this product

Catalog No. AA44940K2



Specifications

Specifications

Tetramethylammonium hydroxide
Colorless to Yellow
75-59-2
C4H13NO
MFCD00008280
3558708
tetramethylammonium hydroxide, tmah, hydroxyde de tetramethylammonium, nmw-w, nmd 3, tetramethyl ammonium hydroxide, unii-5gkp7317q2, ammonium, tetramethyl-, hydroxide, methanaminium, n,n,n-trimethyl-, hydroxide, tetramethylammoniumhydroxide
WGTYBPLFGIVFAS-UHFFFAOYSA-M
tetramethylazanium;hydroxide
60966
Liquid
Electronic
Air Sensitive
1L
(metals basis)
(CH3)4NOH
UN1835
14,9224
2.38% w/w aq. soln.
C[N+](C)(C)C.[OH-]
91.154
91.15
99.9999%

Chemical Identifiers

75-59-2
91.154
WGTYBPLFGIVFAS-UHFFFAOYSA-M
60966
C[N+](C)(C)C.[OH-]
C4H13NO
MFCD00008280
tetramethylammonium hydroxide, tmah, hydroxyde de tetramethylammonium, nmw-w, nmd 3, tetramethyl ammonium hydroxide, unii-5gkp7317q2, ammonium, tetramethyl-, hydroxide, methanaminium, n,n,n-trimethyl-, hydroxide, tetramethylammoniumhydroxide
tetramethylazanium;hydroxide
Safety and Handling

Safety and Handling

GHS H Statement
H300-H310-H370-H372-H314
Fatal if swallowed.
Fatal in contact with skin.
Causes damage to organs.
Causes damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.

GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: TETRAMETHYLAMMONIUM HYDROXIDE SOLUTION

RTECSNumber : PA0875000

TSCA : TSCA

SDS Documents