missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Tetramethylammonium Hydroxide, 10% in Water, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 420511000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalystSpecifications
Tetramethylammonium hydroxide | |
75-59-2,7732-18-5 | |
1.0000g/mL | |
Plastic bottle | |
(CH3)4NOH | |
100 g | |
11,514 | |
15, 9371 | |
Solubility in water: soluble. | |
C[N+](C)(C)C.[OH-] | |
91.15 | |
91.15 | |
Liquid |
10% in Water | |
Colorless | |
100.0°C | |
C4H13NO | |
MFCD00008280 | |
04, 50 | |
1 | |
tetramethylammonium hydroxide, tmah, hydroxyde de tetramethylammonium, nmw-w, nmd 3, tetramethyl ammonium hydroxide, unii-5gkp7317q2, ammonium, tetramethyl-, hydroxide, methanaminium, n,n,n-trimethyl-, hydroxide, tetramethylammoniumhydroxide | |
WGTYBPLFGIVFAS-UHFFFAOYSA-M | |
tetramethylazanium;hydroxide | |
60966 | |
9.5 to 10.5% |
RUO – Research Use Only