Learn More
Tetramethylammonium hydroxide, 1.0 M aq. soln., ACS, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 044938K2
Description
Tetramethylammonium hydroxide is used as anisotropic etching of silicon, as a basic solvent in the development of acidic photo resist in the photolithography process, and is highly effective in stripping photo resist, and is used as a surfactant in the synthesis of ferrofluid, to inhibit nanoparticle aggregation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Tetramethylammonium hydroxide | |
Colorless to Yellow | |
(CH3)4NOH | |
1 L | |
3558708 | |
14,9224 | |
Soluble in water. | |
C[N+](C)(C)C.[OH-] | |
91.154 | |
91.15 | |
1.0M aq. soln. |
75-59-2 | |
C4H13NO | |
MFCD00008280 | |
UN1835 | |
Air Sensitive | |
tetramethylammonium hydroxide, tmah, hydroxyde de tetramethylammonium, nmw-w, nmd 3, tetramethyl ammonium hydroxide, unii-5gkp7317q2, ammonium, tetramethyl-, hydroxide, methanaminium, n,n,n-trimethyl-, hydroxide, tetramethylammoniumhydroxide | |
WGTYBPLFGIVFAS-UHFFFAOYSA-M | |
tetramethylazanium;hydroxide | |
60966 | |
ACS | |
Liquid |
Chemical Identifiers
75-59-2 | |
91.154 | |
WGTYBPLFGIVFAS-UHFFFAOYSA-M | |
60966 | |
C[N+](C)(C)C.[OH-] |
C4H13NO | |
MFCD00008280 | |
tetramethylammonium hydroxide, tmah, hydroxyde de tetramethylammonium, nmw-w, nmd 3, tetramethyl ammonium hydroxide, unii-5gkp7317q2, ammonium, tetramethyl-, hydroxide, methanaminium, n,n,n-trimethyl-, hydroxide, tetramethylammoniumhydroxide | |
tetramethylazanium;hydroxide |
Safety and Handling
GHS H Statement
H301-H311-H314-H371-H373
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
May cause damage to organs.
May cause damage to organs through prolonged or repeated exposure.
P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302-H311-H314-H370-H372
DOTInformation : Hazard Class: 8; Packaging Group: II
EINECSNumber : 200-882-9
RTECSNumber : PA0875000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only