Please login to your online account to display your discounted pricing

Tetramethylammonium hydrogensulfate, 99+%, HPLC grade, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC343830050

 View more versions of this product

Catalog No. AC343830050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥99.0% (on dry substance)
Purity Grade HPLC Grade
Physical Form Powder
Formula Weight 171.21
Quantity 5g
Color White to off-white
Absorbance (0.005 M in water)
Packaging Glass bottle
CAS 80526-82-5
Chemical Name or Material Tetramethylammonium hydrogensulfate
Infrared Spectrum Authentic
Assay Percent Range 99+%
Molecular Formula C4H13NO4S
MDL Number MFCD00036149
Water 1% max. (K.F.)
Absorbance Conditions (0.005M in water)
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Tetramethylammonium hydrogensulfate
CAS 80526-82-5
Purity Grade HPLC Grade
Assay ≥99.0% (on dry substance)
Assay Percent Range 99+%
Molecular Formula C4H13NO4S
Formula Weight 171.21
MDL Number MFCD00036149
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

SDS & Safety Documents