missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetramethylammonium hexafluorophosphate, 99%, ACROS Organics™

Manufacturer:  Acros Organics 138340250

 View more versions of this product

Catalog No. AC138340250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Phase transfer catalyst
Specifications

Specifications

Tetramethylammonium hexafluorophosphate
>300.0°C
558-32-7
Authentic
99%
C4H12F6NP
MFCD00012012
ZFKULDQWLXAKIM-UHFFFAOYSA-N
tetramethylazanium;hexafluorophosphate
11209
Powder
White
25g
98.5% min.
Glass bottle
99%
(CH3)4NPF6
1.4820 to 1.4850
C[N+](C)(C)C.F[P-](F)(F)(F)(F)F
219.111
219.11

Chemical Identifiers

558-32-7
219.111
ZFKULDQWLXAKIM-UHFFFAOYSA-N
C4H12F6NP
MFCD00012012
11209
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS Documents