missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetramethylammonium fluoride tetrahydrate, 98%, ACROS Organics™

Chemical Identifiers

CAS 17787-40-5
Assay Percent Range 98%
Molecular Formula C4H12FN·4H2O
Linear Formula (CH3)4NF·4H2O
Formula Weight 165.21
MDL Number MFCD00149968
Chemical Name or Material Tetramethylammonium fluoride tetrahydrate, 98%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC148070050 SDS Fisher Scientific Acros
148070050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

Phase transfer catalyst
Description & Specifications

Chemical Identifiers

17787-40-5
C4H12FN·4H2O
165.21
Tetramethylammonium fluoride tetrahydrate, 98%
98%
(CH3)4NF·4H2O
MFCD00149968

Specifications

165.21
07,358
(CH3)4NF·4H2O
C4H12FN·4H2O
98%
5g
Glass bottle
MFCD00149968
17787-40-5
Tetramethylammonium fluoride tetrahydrate, 98%

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning