Learn More
Tetramethylammonium chloride, 98+%, Thermo Scientific Chemicals
$76.84 - $119.29
Chemical Identifiers
CAS | 75-57-0 |
---|---|
Molecular Formula | C4H12ClN |
Molecular Weight (g/mol) | 109.6 |
MDL Number | MFCD00011628 |
InChI Key | OKIZCWYLBDKLSU-UHFFFAOYSA-M |
Synonym | tetramethylammonium chloride, tetramethyl ammonium chloride, tetramine chloride, usaf an-8, n,n,n-trimethylmethanaminium chloride, methanaminium, n,n,n-trimethyl-, chloride, unii-dcq9s88703, tetramethylazanium chloride, ammonium, tetramethyl-, chloride, tetramethylammonium chloride tmacl |
PubChem CID | 6379 |
ChEBI | CHEBI:7070 |
IUPAC Name | tetramethylazanium;chloride |
SMILES | C[N+](C)(C)C.[Cl-] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC138331000
|
Thermo Scientific Chemicals
138331000 |
100 g | Plastic bottle |
Each for $76.84
|
|
||||
AC138332500
|
Thermo Scientific Chemicals
138332500 |
250 g | Plastic bottle |
Each for $119.29
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalystChemical Identifiers
75-57-0 | |
109.6 | |
OKIZCWYLBDKLSU-UHFFFAOYSA-M | |
6379 | |
tetramethylazanium;chloride |
C4H12ClN | |
MFCD00011628 | |
tetramethylammonium chloride, tetramethyl ammonium chloride, tetramine chloride, usaf an-8, n,n,n-trimethylmethanaminium chloride, methanaminium, n,n,n-trimethyl-, chloride, unii-dcq9s88703, tetramethylazanium chloride, ammonium, tetramethyl-, chloride, tetramethylammonium chloride tmacl | |
CHEBI:7070 | |
C[N+](C)(C)C.[Cl-] |
Specifications
75-57-0 | |
White to Ivory | |
98% min. (Argentometry) | |
C4H12ClN | |
MFCD00011628 | |
tetramethylammonium chloride, tetramethyl ammonium chloride, tetramine chloride, usaf an-8, n,n,n-trimethylmethanaminium chloride, methanaminium, n,n,n-trimethyl-, chloride, unii-dcq9s88703, tetramethylazanium chloride, ammonium, tetramethyl-, chloride, tetramethylammonium chloride tmacl | |
OKIZCWYLBDKLSU-UHFFFAOYSA-M | |
tetramethylazanium;chloride | |
6379 | |
109.6 | |
98+% | |
Tetramethylammonium chloride, 98+% |
420.0°C | |
Authentic | |
Plastic bottle | |
(CH3)4NCl | |
100 g | |
Solubility in water: >60% (20°C). Other solubilities: insoluble in ether, benzene, slightly soluble in ethanol | |
C[N+](C)(C)C.[Cl-] | |
109.6 | |
CHEBI:7070 | |
Negligible | |
Hygroscopic Crystalline Powder |
Safety and Handling
GHS H Statement
Fatal if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes damage to organs if swallowed.
Toxic to aquatic life with long lasting effects.
GHS P Statement
IF exposed: Call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water
GHS Signal Word: Danger
EINECSNumber : 200-880-8
RTECSNumber : BS7700000
TSCA : TSCA
RUO – Research Use Only