missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Tetramethylammonium chloride, 98+%, ACROS Organics™

Manufacturer:  Acros Organics AC138330051

 View more versions of this product

Catalog No. AC138330051

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Phase transfer catalyst
Specifications

Specifications

Tetramethylammonium chloride
420.0°C
98+%
Vapor Pressure: Negligible
Plastic drum
98+%
(CH3)4NCl
Solubility in water: >60% (20°C). Other solubilities: insoluble in ether, benzene, slightly soluble in ethanol
C[N+](C)(C)C.[Cl-]
109.597
CHEBI:7070
Hygroscopic Crystalline Powder
White through Ivory
5kg
75-57-0
Authentic
≥98%
C4H12ClN
MFCD00011628
OKIZCWYLBDKLSU-UHFFFAOYSA-M
tetramethylazanium;chloride
6379
109.6

Chemical Identifiers

75-57-0
109.597
OKIZCWYLBDKLSU-UHFFFAOYSA-M
CHEBI:7070
C4H12ClN
MFCD00011628
6379
Safety and Handling

Safety and Handling

GHS H Statement
Fatal if swallowed. Toxic in contact with skin. Causes skin irritation. Causes damage to organs if swallowed. Toxic to aquatic life with long lasting effects.

GHS P Statement
IF exposed: Call a POISON CENTER or doctor/physician. Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. Do not breathe dust/fume/gas/mist/vapors/spray. Take off contaminated clothing and wash before reuse. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Danger

EINECSNumber : 200-880-8

RTECSNumber : BS7700000

TSCA : TSCA

SDS Documents