Learn More
Tetramethylammonium bromide, 98%, Thermo Scientific Chemicals
$66.11 - $727.84
Chemical Identifiers
CAS | 64-20-0 |
---|---|
Molecular Formula | C4H12BrN |
Molecular Weight (g/mol) | 154.05 |
MDL Number | MFCD00011626 |
InChI Key | DDFYFBUWEBINLX-UHFFFAOYSA-M |
Synonym | tetramethylammonium bromide, tetramethylammoniumbromide, tetramethylazanium bromide, methanaminium, n,n,n-trimethyl-, bromide, n,n,n-trimethylmethanaminium bromide, unii-6l9ne9609p, ammonium, tetramethyl-, bromide, methanaminium, n,n,n-trimethyl-, bromide 1:1, c4h12n.br, acmc-209nkm |
PubChem CID | 66137 |
ChEBI | CHEBI:55317 |
IUPAC Name | tetramethylazanium;bromide |
SMILES | [Br-].C[N+](C)(C)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1336822
|
Thermo Scientific Chemicals
A1336822 |
100 g |
Each for $66.11
|
|
|||||
AAA1336836
|
Thermo Scientific Chemicals
A1336836 |
500 g |
Each for $204.53
|
|
|||||
AAA133680E
|
Thermo Scientific Chemicals
A133680E |
2500 g |
Each for $727.84
|
|
|||||
Description
Tetramethylammonium bromide acts as an ion pair reagent. It is used as a surface-active agent and an active ingredient for conditioners. Further, it is used as a softner for textiles and paper products. It is also employed as a disinfection agent, sanitizer, antimicrobial and antistatic agent. In addition to this, it plays in important role as a phase transfer catalyst in synthetic chemistry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTetramethylammonium bromide acts as an ion pair reagent. It is used as a surface-active agent and an active ingredient for conditioners. Further, it is used as a softner for textiles and paper products. It is also employed as a disinfection agent, sanitizer, antimicrobial and antistatic agent. In addition to this, it plays in important role as a phase transfer catalyst in synthetic chemistry.
Solubility
Soluble in water, alcohol, chloroform and acetone.
Notes
Hygroscopic. Incompatible with oxidizing agents and strong bases.
Chemical Identifiers
64-20-0 | |
154.05 | |
DDFYFBUWEBINLX-UHFFFAOYSA-M | |
66137 | |
tetramethylazanium;bromide |
C4H12BrN | |
MFCD00011626 | |
tetramethylammonium bromide, tetramethylammoniumbromide, tetramethylazanium bromide, methanaminium, n,n,n-trimethyl-, bromide, n,n,n-trimethylmethanaminium bromide, unii-6l9ne9609p, ammonium, tetramethyl-, bromide, methanaminium, n,n,n-trimethyl-, bromide 1:1, c4h12n.br, acmc-209nkm | |
CHEBI:55317 | |
[Br-].C[N+](C)(C)C |
Specifications
64-20-0 | |
1.56 | |
C4H12BrN | |
MFCD00011626 | |
UN2811 | |
Hygroscopic | |
Soluble in water,alcohol,chloroform and acetone. | |
[Br-].C[N+](C)(C)C | |
154.05 | |
CHEBI:55317 | |
98% |
>300°C | |
Odorless | |
(CH3)4NBr | |
100 g | |
3620955 | |
tetramethylammonium bromide, tetramethylammoniumbromide, tetramethylazanium bromide, methanaminium, n,n,n-trimethyl-, bromide, n,n,n-trimethylmethanaminium bromide, unii-6l9ne9609p, ammonium, tetramethyl-, bromide, methanaminium, n,n,n-trimethyl-, bromide 1:1, c4h12n.br, acmc-209nkm | |
DDFYFBUWEBINLX-UHFFFAOYSA-M | |
tetramethylazanium;bromide | |
66137 | |
154.06 | |
Tetramethylammonium bromide |
Safety and Handling
GHS H Statement
H300-H315-H319-H335
Fatal if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H300-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 200-581-2
RTECSNumber : BS7600000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only