Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetramethylammonium borohydride, 95%

$44.45 - $44.45

Chemical Identifiers

Chemical Name or Material Tetramethylammonium borohydride
CAS 16883-45-7
Molecular Formula C4H16BN
Linear Formula (CH3)4NBH4
Formula Weight 88.99
MDL Number MFCD00011778
UN Number UN3131
Synonym Tetramethylammonium tetrahydridoborate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1765506

  SDS
alfa aesar™
A1765506
5g Each for $44.45

AAA1765522

  SDS
alfa aesar™
A1765522
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 88.99
MDL Number MFCD00011778
Quantity 5g
Assay 95%
Melting Point >250°C (dec.)
Health Hazard 2 GHS H Statement
H261-H301-H311-H314-H318
In contact with water releases flammable gas.
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Chemical Name or Material Tetramethylammonium borohydride
Linear Formula (CH3)4NBH4
Health Hazard 1 Danger
RTECS Number BS8310000
Odor Odorless
DOT Information Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: WATER-REACTIVE SOLID, CORROSIVE, N.O.S.
Synonym Tetramethylammonium tetrahydridoborate
Molecular Formula C4H16BN
UN Number UN3131
TSCA TSCA
CAS 16883-45-7
Health Hazard 3 GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P310-P370+P378i-P402+P404
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
In case of fire: Use for extinction: Fire-extinguishing powder.
Store in a dry place.
Store in a closed container.
Safety and Handling

Danger

GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P310-P370+P378i-P402+P404
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
In case of fire: Use for extinction: Fire-extinguishing powder.
Store in a dry place.
Store in a closed container.

GHS H Statement
H261-H301-H311-H314-H318
In contact with water releases flammable gas.
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.