Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetramethylammonium borohydride, 95%

Chemical Identifiers

Chemical Name or Material Tetramethylammonium borohydride
CAS 16883-45-7
Formula Weight 88.99g/mol
MDL Number MFCD00011778
UN Number UN1409
Synonym Tetramethylammonium borohydride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3257404

 
alfa aesar™
32574-04
2g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Density 0.813g/mL
Melting Point 150°C (dec.) (vac.)
Quantity 2g
Chemical Name or Material Tetramethylammonium borohydride
CAS 16883-45-7
Physical Form Crystalline
Assay 95%
Formula Weight 88.99g/mol
MDL Number MFCD00011778
UN Number UN1409
Synonym Tetramethylammonium borohydride
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H260-H300-H311-H315-H319-H335
In contact with water releases flammable gases which may ignite spontaneously.
Fatal if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P231+P232-P301+P310-P305+P351+P338-P361-P405-P501a
Handle under inert gas.
Protect from moisture.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: METAL HYDRIDES, WATER REACTIVE, N.O.S.
TSCA TSCA
Safety and Handling

Danger

GHS P Statement
P231+P232-P301+P310-P305+P351+P338-P361-P405-P501a
Handle under inert gas.
Protect from moisture.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H260-H300-H311-H315-H319-H335
In contact with water releases flammable gases which may ignite spontaneously.
Fatal if swallowed.
Toxic in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.