Please login to your online account to display your discounted pricing

Alfa Aesar™ Tetramethylammonium bis(trifluoromethylsulfonyl)imide, 97%

Chemical Identifiers

Chemical Name or Material Tetramethylammonium bis(trifluoromethylsulfonyl)imide
CAS 161401-25-8
Linear Formula (CF3SO2)2NN(CH3)4
Formula Weight 354.28
MDL Number MFCD23098729
UN Number UN2923
Synonym Tetramethylammonium triflimide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4700403

 
alfa aesar™
4700403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4700406

 
alfa aesar™
4700406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 354.28
Melting Point 132° to 136°C
Chemical Name or Material Tetramethylammonium bis(trifluoromethylsulfonyl)imide
Physical Form Powder
Assay 97%
Linear Formula (CF3SO2)2NN(CH3)4
MDL Number MFCD23098729
CAS 161401-25-8
UN Number UN2923
Synonym Tetramethylammonium triflimide
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H311-H314-H318
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Storage Note 1 2° to 8°C
DOT Information Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
Sensitivity Moisture Sensitive
Color White
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H311-H314-H318
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.

2° to 8°C